Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipin 3 Antibody (OTI1D7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272160FR
Description
Lipin 3 Monoclonal specifically detects Lipin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lipin 3 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
LPIN3 | |
Full length recombinant protein of human Lipin 3 (NP_075047) produced in HEK293T cells. | |
0.1 mL | |
64900.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1D7 | |
Western Blot, Immunohistochemistry | |
dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, phosphatidate phosphatase LPIN3, SMP2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction