Learn More
Invitrogen™ LIPK Polyclonal Antibody

Description
LIPK Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | LIPK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LIPK |
| Gene Accession No. | Q5VXJ0 |
| Gene Alias | 9930115F20; bA186O14.2; BC055815; lipase family member K; lipase K; lipase member K; lipase, family member K; lipase-like abhydrolase domain-containing protein 2; lipase-like, ab-hydrolase domain containing 2; Lipk; Lipl2 |
| Gene Symbols | LIPK |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.