Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIPK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989283
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9283 Supplier Bioss Supplier No. BS9711R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Plays a highly specific role in the last step of keratinocyte differentiation. May have an essential function in lipid metabolism of the most differentiated epidermal layers.

Specifications

Antigen LIPK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LIPK
Gene Accession No. Q5VXJ0
Gene Alias bA186O14.2; lipase family member K; lipase K; Lipase member K; lipase, family member K; Lipase-like abhydrolase domain-containing protein 2; lipase-like, ab-hydrolase domain containing 2; LIPK; LIPL2
Gene Symbols LIPK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LIPK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643414
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.