Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIPM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145001
Description
Immunogen sequence: SVHMPTKAVD PEAFMNISEI IQHQGYPCEE YEVATE Antigen sequence identity: Mouse 81%; Rat 83%.
Plays a highly specific role in the last step of keratinocyte differentiation. May have an essential function in lipid metabolism of the most differentiated epidermal layers.
Specifications
LIPM | |
Polyclonal | |
Unconjugated | |
LIPM | |
bA304I5.1; lipase family member M; lipase M; lipase member M; lipase, family member M; lipase-like abhydrolase domain-containing protein 3; lipase-like, ab-hydrolase domain containing 3; LIPL3; LIPM | |
Rabbit | |
Affinity chromatography | |
RUO | |
340654 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VYY2 | |
LIPM | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 33-68. Recombinant protein control fragment (Product #RP-109853). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction