Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571721
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571721 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571721 Supplier Invitrogen™ Supplier No. PA571721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIPM Polyclonal Antibody for Western Blot, IHC (P)

Plays a highly specific role in the last step of keratinocyte differentiation. May have an essential function in lipid metabolism of the most differentiated epidermal layers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LIPM
Gene Accession No. Q5VYY2
Gene Alias bA304I5.1; lipase family member M; lipase M; lipase member M; lipase, family member M; lipase-like abhydrolase domain-containing protein 3; lipase-like, ab-hydrolase domain containing 3; LIPL3; LIPM
Gene Symbols LIPM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 318-346 amino acids from the C-terminal region of human LIPM.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.