Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545806 Supplier Invitrogen™ Supplier No. PA545806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IFTRFFLLPN SIIKAVFGTK GFFLEDKKTK IASTKICNNK ILWLICSEFM Sequence homology: Human: 100%.

The gene encodes a lipase that is highly expressed in granular keratinocytes in the epidermis, and plays a role in the differentiation of keratinocytes. Mutations in this gene are associated with lamellar ichthyosis type 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPN
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIPN
Gene Accession No. Q5VXI9
Gene Alias ARCI8; bA186O14.3; LI4; lipase family member N; lipase member N; lipase, family member N; lipase-like abhydrolase domain-containing protein 4; LIPL4; LIPN
Gene Symbols LIPN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human LIPN (aa 211-260).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643418
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.