Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299834PCP
Description
Lipocalin-1 Polyclonal antibody specifically detects Lipocalin-1 in Human samples. It is validated for Western Blot, ELISASpecifications
Lipocalin-1 | |
Polyclonal | |
Western Blot, ELISA | |
lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin) | |
Produced in rabbits immunized with purified, recombinant Human Lipocalin-1 (Accession#: NP_002288.1; Met 1-Asp176) | |
0.1 mL | |
Cancer | |
3933 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction