Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-2/NGAL Antibody (128), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289346MFV450
Description
Lipocalin-2/NGAL Monoclonal antibody specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, ELISASpecifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 128 | |
| Western Blot, ELISA | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Lipocalin-2/NGAL (Accession#: NP_005555.2; Met 1-Gly 198). | |
| 0.1 mL | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 3934 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction