Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipocalin-2/NGAL Antibody (14), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP241365AF594

Catalog No. NP24136A94


Only null left
Add to Cart

Description

Description

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for ELISA.
Specifications

Specifications

Lipocalin-2/NGAL
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
LCN2
Human recombinant NGAL coupled to carrier.
100 μL
Primary
Antibody is specific for human NGAL (neutrophil gelatinase-associated lipocalin).
Store at 4°C in the dark.
ELISA
14
ELISA
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
Mouse
Protein A or G purified
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience
3934.0
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.