Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-2/NGAL Antibody (14), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241364AF594
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Canine, Human (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA.Specifications
Lipocalin-2/NGAL | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
LCN2 | |
Recombinant dog NGAL adsorbed onto aluminum hydroxide gel | |
100 μL | |
Primary | |
Antibody binds dog NGAL | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry | |
14 | |
Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA | |
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
Mouse | |
Protein A or G purified | |
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
3934.0 | |
Canine, Human, Mouse, Rat | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction