Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-2/NGAL Antibody (29) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$243.50 - $560.50
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Clone | 29 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Lipocalin-2/NGAL | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 3934 | |
| Recombinant mouse NGAL adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4C. Do not freeze. |
| 29 | |
| Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| LCN2 | |
| IgG1 κ | |
| Protein A or G purified | |
| neutrophil gelatinase-associated lipocalin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title