Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipocalin-2/NGAL Rabbit anti-Mouse, Clone: 211, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $464.00
Specifications
Antigen | Lipocalin-2/NGAL |
---|---|
Clone | 211 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for ELISA.Specifications
Lipocalin-2/NGAL | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
Recombinant Mouse Lipocalin-2/NGAL protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
211 | |
ELISA | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
0.2 um filtered solution in PBS | |
3934 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title