Learn More
Description
Designed for high-content screening (HCS) workflows, it offers exceptional signal-to-noise ratios and photostability, making it the ideal partner for metabolic dysfunction-associated steatohepatitis (MASH), neurodegeneration, and oncology studies (demonstrated staining of U2OS and iPSC derived neurons). It is provided as 50 μL of a 1000? concentrate in DMSO. Excitation and emission maxima (λ) are 488 nm and 525-550 nm, respectively.
LipoLIVE™ is a trademark of Saguaro Biosciences.
Specifications
Specifications
| Quantity | 50 uL |
| Product Line | Tocris |
| Target | Live-cell Imaging |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
