Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipoprotein Lipase/LPL Antibody (OTI3A10), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271178F

Catalog No. NBP271178F


Only null left
Add to Cart

Description

Description

Lipoprotein Lipase/LPL Monoclonal specifically detects Lipoprotein Lipase/LPL in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Lipoprotein Lipase/LPL
Monoclonal
FITC
PBS with 0.05% Sodium Azide
LPL
Recombinant protein fragment corresponding to amino acids 28-475 of human Lipoprotein Lipase/LPL (NP_000228) produced in HEK293T.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry, Immunohistochemistry
OTI3A10
Flow Cytometry, Immunohistochemistry
EC 3.1.1, HDLCQ11, LIPDEC 3.1.1.34, lipoprotein lipase
Mouse
Affinity Purified
Lipid and Metabolism
4023.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.