Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lipoprotein a Native Protein

Click to view available options
:
0.05mg; Unlabeled
Description
Native Lipoprotein a The Lipoprotein a Native Protein is derived from Human Plasma. The Lipoprotein a Native Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Accession Number | P08519 |
Formulation | Liquid in 150mM NaCl, pH 7.4 with 0.01% EDTA |
Gene ID (Entrez) | 4018 |
Molecular Weight (g/mol) | 280-700kDa |
Name | Lipoprotein a Protein |
Purification Method | SDS-PAGE |
Storage Requirements | 4°C. Do not freeze. |
Purity or Quality Grade | >95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction