Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Liprin alpha 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Liprin alpha 1 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Liprin alpha 1 | |
Polyclonal | |
Rabbit | |
Human | |
Q13136 | |
8500 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QLEETQHDKDQLVLNIEALRAELDHMRLRGASLHHGRPHLGSVPDFRFPMADGHTDSYSTSAVLRRPQKGRLAALRDEPSKVQTLNEQDWERAQQASVLANVAQAFESDADVSDGEDDRDTLLSSVDLLS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
PPFIA1 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title