Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Liprin alpha 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215018
Description
Liprin alpha 2 Polyclonal specifically detects Liprin alpha 2 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Liprin alpha 2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
PPFIA2 | |
Recombinant protein encompassing a sequence within the center region of human Liprin alpha 2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
8499 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.98mg/mL | |
Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Protein tyrosine phosphatase receptor type f polypeptide interacting protein alpha 2, PTPRF interacting protein alpha 2 | |
Rabbit | |
143 kDa | |
0.1 mL | |
Primary | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction