Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPT2-AS1 Antibody (OTI4C7), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271949AF405
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytochrome c | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CYCS | |
Full length recombinant protein of human Cytochrome c (NP_940921) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4C7 | |
Western Blot, Immunohistochemistry | |
CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
Mouse | |
Affinity Purified | |
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
54205.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction