Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPT2-AS1 Antibody (OTI4C7), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271949AF488
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Cytochrome c | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYCS | |
| Full length recombinant protein of human Cytochrome c (NP_940921) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4C7 | |
| Western Blot, Immunohistochemistry | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction