Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPT2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548972
Description
Catalyzes the transfer of endogenously produced octanoic acid from octanoyl-acyl-carrier-protein onto the lipoyl domains of lipoate-dependent enzymes. Lipoyl-ACP can also act as a substrate although octanoyl-ACP is likely to be the physiological substrate.
Specifications
LIPT2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
A6NK58 | |
LIPT2 | |
KLH conjugated synthetic peptide between 13-43 amino acids from the N-terminal region of human LIPT2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
LIPT2 | |
Putative lipoyltransferase 2, mitochondrial, Lipoate-protein ligase B, Lipoyl/octanoyl transferase, Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase, LIPT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
387787 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction