Learn More
Liquid Accelerometer Kit
Click to view available options
:
Liquid Accelerometer Kit
Description
Includes
Hook and loop connector to attach to a dynamics cart or rotating platform; grid lines to quantify both vertical and horizontal movement; instructions
Requires
Water, food coloring
Horizontal acceleration creates characteristic wave patterns to demonstrate inertial movement.
- See these waves by watching the pattern created by the colored liquid
- Measure the patterns with the imprinted horizontal and vertical grid
- Demonstrate gravitational and uniform acceleration Illustrate acceleration down an inclined plane Use with dynamics carts, rotating platform, turntable or amusement park physics
Specifications
Specifications
Product Type | Kit, Liquid Accelerometer |
For Use With (Application) | Demonstrate inertial movement |
Includes | Hook and loop connector to attach to a dynamics cart or rotating platform, grid lines to quantify both vertical and horizontal movement, instructions |