Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Cell Company Liquid Filter Set for Testing Photometric Accuracy in UV/VIS Range

Catalog No. 14397525
Click to view available options
Type:
Liquid Filter Set

Includes

UV60: Absorption filter (0.75 Abs), UV600: Absorption filter (1.0 Abs), UV14: Reference filter, DAkkS calibration certificate

For testing photometric accuracy in UV and Vis range, according to European Pharmacopoeia.

  • Certified at wavelengths: 235, 257, 313, 350, 430nm
  • Slit width: 2nm
  • Supplied in high-quality storage case

Specifications

Type Liquid Filter Set
For Use With (Application) Testing photometric accuracy in UV and Vis range, according to European Pharmacopoeia.
Includes UV60: Absorption filter (0.75 Abs), UV600: Absorption filter (1.0 Abs), UV14: Reference filter, DAkkS calibration certificate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.