Learn More
Cliniqa™ Liquid QC™ CoV-2 IgG Control
Description
Clinqa™ Liquid QC™ CoV-2 IgG Control Kit is a liquid stable, ready to use bi-level control for use with assays designed to produce qualitative results for Covid-19 IgG antibodies to the SARS-CoV-2 spike (S) protein. Intended for in vitro diagnostic use.
The Liquid QC™ CoV-2 IgG Control kit provides two distinct levels. Each kit includes six 3 mL filled vials per level. All laboratories should establish their own expected ranges and tolerance limits based on their test system.
Compatible with the following instruments and assays:
- Ortho Vitros™ Anti-SARS-CoV-2 IgG Test
- Beckman Coulter Access™ SARS-CoV-2 IgG Antibody Test
- Siemens™ SARS -CoV-2 Total Antibody Test
- Biomerieux™ VIDAS™ anti-SARS-CoV-2 IgG Test
Specifications
Specifications
Detectable Analytes | IgG |
Detection Method | Control |
For Use With (Equipment) | Ortho Vitros™, Beckman Coulter Access™, Siemens Dimension™, Biomerieux™ Vidas™ Antibody Tests |
Sample Type | Whole Blood, Serum, Plasma |
Type | Vials |
Quantity | 12 Vials |
Stability | 30 Days at 2°C to 8°C |
Form | Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.