Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Liquified Phenol

Catalog No. P9346100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P9346100ML 100 g
P9346500ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. P9346100ML Supplier Sigma Aldrich Supplier No. P9346100ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Liquified Phenol is a solution of phenol containing ~10% water. It is potential as a chemical peeling agent for treating stable vitiligo and alopecia areata.Liquified Phenol has been used in a study to record the one-photon (linear) absorption (1PA) and two-photon absorption 2PA spectra of Disperse Red One (DR1). It may be used for deproteinization of cell lysate during the preparation of total nucleic acids.

Specifications

CAS 9003-39-8
Linear Formula (C6H9NO)n
MDL Number MFCD00149016
Quantity 100 g
Synonym PVP; Polyvidone; Povidone
Molecular Weight (g/mol) 10000 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.