Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Liquified Phenol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 g
100 mL
Description
Liquified Phenol is a solution of phenol containing ~10% water. It is potential as a chemical peeling agent for treating stable vitiligo and alopecia areata.Liquified Phenol has been used in a study to record the one-photon (linear) absorption (1PA) and two-photon absorption 2PA spectra of Disperse Red One (DR1). It may be used for deproteinization of cell lysate during the preparation of total nucleic acids.
Specifications
Specifications
| CAS | 108-95-2 |
| Density | 1.07 g/mL at 25°C |
| Molecular Formula | C6H6O |
| Linear Formula | C6H6O |
| Quantity | 100 mL |
| MDL Number | MFCD00002143 |
| Synonym | Phenol solution |
| Molecular Weight (g/mol) | 94.11 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction