Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIS1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200326C
Description
LIS1 Polyclonal antibody specifically detects LIS1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
LIS1 | |
Polyclonal | |
Western Blot | |
LIS-1, LIS1PAF-AH alpha, LIS2, lissencephaly 1 protein, Lissencephaly-1 protein, MDCR, MDS, Miller-Dieker syndrome chromosome region, PAF acetylhydrolase 45 kDa subunit, PAFAH, PAF-AH 45 kDa subunit, PAFAH alpha, PAFAHA, platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa), platelet-activating factor acetylhydrolase IB subunit alpha, platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD), platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa) | |
Rabbits were immunized with a synthetic peptide, which represented a portion of human Lissencephaly-1 protein encoded in part by exons 3 and 4 (LocusLink ID 5048). Antibody was affinity purified using the peptide immobilized on solid support. | |
0.1 mL | |
Cell Cycle and Replication | |
5048 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction