Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP187769
Description
LIS1 Polyclonal specifically detects LIS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LIS1 | |
Polyclonal | |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
LIS-1, LIS1PAF-AH alpha, LIS2, lissencephaly 1 protein, Lissencephaly-1 protein, MDCR, MDS, Miller-Dieker syndrome chromosome region, PAF acetylhydrolase 45 kDa subunit, PAFAH, PAF-AH 45 kDa subunit, PAFAH alpha, PAFAHA, platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa), platelet-activating factor acetylhydrolase IB subunit alpha, platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD), platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PAFAH1B1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ADYLRSNGYEEAYSVFKKEAELDVNEELDKKYAGLLEKKWTSVIRLQKKVMELESKLNEAKEEFTSGGPLGQKRDPKEWIPRPPEKYALSGHRSPVTRVIFHPVFSVMVSASEDATIKVWDYETGDFERTLKGHTDSVQDISFDHS | |
0.1 mL | |
Cell Cycle and Replication | |
5048 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction