Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIS1 Rabbit anti-Human, Mouse, Rat, Clone: 9O6X3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | LIS1 |
---|---|
Clone | 9O6X3 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
LIS1 Monoclonal antibody specifically detects LIS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
LIS1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5048 | |
IgG | |
Affinity purified |
9O6X3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
LIS-1, LIS1PAF-AH alpha, LIS2, lissencephaly 1 protein, Lissencephaly-1 protein, MDCR, MDS, Miller-Dieker syndrome chromosome region, PAF acetylhydrolase 45 kDa subunit, PAFAH, PAF-AH 45 kDa subunit, PAFAH alpha, PAFAHA, platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa), platelet-activating factor acetylhydrolase IB subunit alpha, platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD), platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa) | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LIS1 (P43034). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title