Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIS1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984450
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4450 Supplier Bioss Supplier No. BS2035R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.

Specifications

Antigen LIS1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PAFAH1B1
Gene Accession No. P43034
Gene Alias BOS_18531; HAMAP-Rule:MF_03141}; LIS1; Lis-1; LIS-1 {ECO:0000255; LIS2; lissencephaly 1 protein; Lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255; LOC100354130; MDCR; MDS; Mdsh; MGC25297; Miller-Dieker syndrome chromosome region; MMS10-U; Ms10u; NudF; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255; PAFAH; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255; PAFAH alpha; PAF-AH alpha; PAFAH alpha {ECO:0000255; PAF-AH alpha {ECO:0000255; PAF-AH beta; Pafah1b1; Pafaha; platelet activating factor acetylhydrolase 1b regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); Platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa); RP23-194P5.2
Gene Symbols PAFAH1B1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LIS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5048
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.