Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria Monocytogenes Antibody (LZH1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11010377
Description
Listeria Monocytogenes Monoclonal specifically detects Listeria Monocytogenes in Bacteria samples. It is validated for Western Blot, Sandwich ELISA Capture.Specifications
Listeria Monocytogenes | |
Monoclonal | |
Unconjugated | |
L. monocytogenes | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified L. monocytogenes outer membrane fraction (OM). | |
0.2 mg | |
Virology Bacteria and Parasites | |
Reacts with L. Monocytogenes outer membrane fraction and intact cells. | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, ELISA | |
LZH1 | |
Western Blot 1:100-1:2000, Sandwich ELISA Capture 1:100-1:2000 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction