Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Listeria monocytogenes Cornell University derived from Silliker SLR2249
Shop All Microbiologics Products

Click to view available options
Quantity:
2 KWIK-STIK
Description
- Biosafety Level: 2
- 2 self-contained units of a single organism
- Act A gene removed
Specifications
Specifications
ATCC® | Silliker SLR2249 |
Biosafety Level | 2 |
Content And Storage | 2°C to 8°C |
Format | KWIK-STIK 2 Pack |
Manufacturing Origin | United States |
Product Type | KWIK-STIK |
For Use With (Application) | Food and Food Ingredients: Pathogen Testing, Microbial Identification |
Packaging Type | Foil Sleeves |
Quantity | 2 KWIK-STIK |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction