Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Listeria monocytogenes Monoclonal Antibody (4212)
GREENER_CHOICE

Catalog No. PIMA533271
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533271 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533271 Supplier Invitrogen™ Supplier No. MA533271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pairs as the conjugate with MA5-33270.

The genus Listeria comprises six species: L. monocytogenes, L. innocua, L. welshimeri, L. seeligeri, L. ivanovii and L. grayi. Listeria monocytogenes, the most commonly isolated pathogenic member, is associated with a wide spectrum of human and animal diseases. In the smear from the original tissue, L. monocytogenes may appear as gram-positive coccobacilli that may be confused with Streptococcus agalactiae (group B), enterococci, or Corynebacterium spp. Listeria is differentiated from streptococci by a positive catalase test.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Listeria monocytogenes
Applications ELISA
Classification Monoclonal
Clone 4212
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias L. monocytogenes
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.