Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria Monocytogenes Antibody (LZF7), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001281IR
Description
Listeria Monocytogenes Monoclonal specifically detects Listeria Monocytogenes in Bacteria samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
Listeria Monocytogenes | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |
Western Blot, ELISA | |
LZF7 | |
Western Blot, ELISA, Sandwich ELISA | |
L. monocytogenes | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified L. monocytogenes outer membrane fraction (OM). | |
0.2 mL | |
Virology Bacteria and Parasites | |
MAbs recognize Listeria monocytogenes outer membrane fraction and intact L. monocytogenes cells. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction