Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Listeria monocytogenes p60 Antibody (p6017), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280121B
This item is not returnable.
View return policy
Description
Listeria monocytogenes p60 Monoclonal antibody specifically detects Listeria monocytogenes p60 in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Listeria monocytogenes p60 | |
Monoclonal | |
Biotin | |
IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein | |
Recombinant Listeria monocytogenes p60. | |
0.1 mL | |
Immunology | |
Bacteria | |
Purified |
Western Blot, ELISA | |
p6017 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction