Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Listeria monocytogenes Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA17230

Catalog No. PA17230


Only null left
Add to Cart

Description

Description

PA1-7230 detects Listeria monocytogenes. PA1-7230 has been successfully used in ELISA procedures. The PA1-7230 immunogen is Listeria monocytogenes; ATCC strain 43251. Recommended short term storage of less than 6 months at 2-8ºC. For extended storage, aliquot and freeze.

The genus Listeria comprises six species: L. monocytogenes, L. innocua, L. welshimeri, L. seeligeri, L. ivanovii and L. grayi. Listeria monocytogenes, the most commonly isolated pathogenic member, is associated with a wide spectrum of human and animal diseases. In the smear from the original tissue, L. monocytogenes may appear as gram-positive coccobacilli that may be confused with Streptococcus agalactiae (group B), enterococci, or Corynebacterium spp. Listeria is differentiated from streptococci by a positive catalase test.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Listeria monocytogenes
Polyclonal
Unconjugated
L. monocytogenes
Listeria monocytogenes; ATCC strain 43251.
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
Protein A
RUO
Bacteria
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.