Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LITAF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102778 Supplier Invitrogen™ Supplier No. PA5102778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LITAF.

Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LITAF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LITAF
Gene Accession No. P0C0T0, Q99732, Q9JLJ0
Gene Alias 3222402J11Rik; C85531; CMT1C; EET-1; Estrogen-enhanced transcript protein; estrogen-enhanced transcript protein 1; estrogen-responsive uterine transcript; FLJ38636; lipopolysaccharide induced TNF factor; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; lipopolysaccharide-induced tumor necrosis factor-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor homolog; Litaf; LITAF-like protein; LPS-induced TN factor; LPS-induced TNF-alpha factor; LPS-induced TNF-alpha factor homolog; mEET; MGC116698; MGC116700; MGC116701; MGC125274; MGC125275; MGC125276; N4wbp3; Nedd4 WW domain-binding protein 3; p53-induced gene 7 protein; PIG 7; PIG7; SIMPLE; Small integral membrane protein of lysosome/late endosome; Tbx1; TP53I7; tumor protein p53 inducible protein 7
Gene Symbols LITAF
Host Species Rabbit
Immunogen A synthesized peptide derived from human LITAF(Accession Q99732), corresponding to amino acid residues S31-V81.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56722, 65161, 9516
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.