Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LITAF Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517242
Description
It is not recommended to aliquot this antibody.
Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.
Specifications
| LITAF | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q99732 | |
| LITAF | |
| Synthetic peptide corresponding to residues near the amino terminus of human LITAF protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LITAF | |
| 3222402J11Rik; C85531; CMT1C; EET-1; Estrogen-enhanced transcript protein; estrogen-enhanced transcript protein 1; estrogen-responsive uterine transcript; FLJ38636; lipopolysaccharide induced TNF factor; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; lipopolysaccharide-induced tumor necrosis factor-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor homolog; Litaf; LITAF-like protein; LPS-induced TN factor; LPS-induced TNF-alpha factor; LPS-induced TNF-alpha factor homolog; mEET; MGC116698; MGC116700; MGC116701; MGC125274; MGC125275; MGC125276; N4wbp3; Nedd4 WW domain-binding protein 3; p53-induced gene 7 protein; PIG 7; PIG7; SIMPLE; Small integral membrane protein of lysosome/late endosome; Tbx1; TP53I7; tumor protein p53 inducible protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9516 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction