Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LITAF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552367 Supplier Invitrogen™ Supplier No. PA552367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APPSYEETVA VNSYYPTPPA PMPGPTTGLV TGPDGKGMNP PSYYTQPAPI PNNNPITVQT VYVQHPITFL DRPIQMCCPS CNKMIVSQLS YNAGALTWLS CGSLCLLGCI AGCCFIPFCV DALQDVDHYC PNC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LITAF
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LITAF
Gene Accession No. Q99732
Gene Alias 3222402J11Rik; C85531; CMT1C; EET-1; Estrogen-enhanced transcript protein; estrogen-enhanced transcript protein 1; estrogen-responsive uterine transcript; FLJ38636; lipopolysaccharide induced TNF factor; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; lipopolysaccharide-induced tumor necrosis factor-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor homolog; Litaf; LITAF-like protein; LPS-induced TN factor; LPS-induced TNF-alpha factor; LPS-induced TNF-alpha factor homolog; mEET; MGC116698; MGC116700; MGC116701; MGC125274; MGC125275; MGC125276; N4wbp3; Nedd4 WW domain-binding protein 3; p53-induced gene 7 protein; PIG 7; PIG7; SIMPLE; Small integral membrane protein of lysosome/late endosome; Tbx1; TP53I7; tumor protein p53 inducible protein 7
Gene Symbols LITAF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LITAF. Recombinant protein control fragment (Product #RP-89429).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9516
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.