Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 2,2-di-n-propylacetate, 97+%, Thermo Scientific™

Catalog No. AA4479809
Click to view available options
Quantity:
0.5 g
2 g
10 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93391-29-8
Molecular Formula C8H15LiO2
Molecular Weight (g/mol) 150.15
MDL Number MFCD04113112
InChI Key CWFAFDCTBAYTFA-UHFFFAOYSA-M
Synonym valproic acid lithium
PubChem CID 23716278
IUPAC Name lithium;2-propylpentanoate
SMILES [Li+].CCCC(CCC)C([O-])=O

Specifications

Quantity 10 g
Formula Weight 150.14
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Lithium 2,2-di-n-propylacetate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.