Learn More
Lithium acetate dihydrate, Reagent Grade
Description
Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 54°C to 57°C |
Color | White |
Physical Form | Crystals |
Quantity | 1 kg |
Beilstein | 3564320 |
Merck Index | 14,5521 |
Solubility Information | Freely soluble in water and alcoholSoluble in water and ethanol. |
Formula Weight | 102.01 (65.98 Anhydrous) |
Grade | Reagent |
Density | 1.300 g/mL |
Show More |
Safety and Handling
Hazard Category | H319 |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 208-914-3 |
RTECSNumber | AI5450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.