Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium acetate dihydrate, Reagent Grade

Catalog No. p-7049225
Click to view available options
Quantity:
250 g
1 kg

CAS: 6108-17-4 | C2H7LiO4 | 102.014 g/mol

Lithium acetate dihydrate is used as a buffer component in the modified ninhydrin reagent for amino analysis by ion-exchange chromatography. It is used as a catalyst in organic reactions, alkyd resin and acrylic polymer production and as an anticorrosion agent in molding polyphenylene sulfide resins as well as in the separation of saturated and unsaturated fatty acids. In the pharmaceutical industry, it is utilized for the preparation of diuretics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.014
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 54°C to 57°C
Physical Form Crystals
Quantity 250 g
Beilstein 3564320
Merck Index 14,5521
Solubility Information Freely soluble in water and alcoholSoluble in water and ethanol.
Formula Weight 102.01 (65.98 Anhydrous)
Grade Reagent
Density 1.3 g/mL
Chemical Name or Material Lithium acetate dihydrate
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 208-914-3
RTECSNumber AI5450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.