Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium aluminum hydride, 15% solution in toluene/THF, Thermo Scientific™
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagent
Specifications
Specifications
| Color | Turbid solution |
| Linear Formula | LiAlH4 |
| Concentration | ca 15%, exact strength on the certificate of analysis |
| Density | 0.8900g/mL |
| Formula Weight | 37.95 |
| Packaging | AcroSeal™ Glass bottle |
| Specific Gravity | 0.89 |
| Quantity | 100 mL |
| Flash Point | −20°C |
| Chemical Name or Material | Lithium Aluminum hydride, 15% solution in toluene/THF |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Suspected of damaging the unborn child. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. May cause drowsiness or dizziness. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction