Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminum hydride, 4.0M solution in diethyl ether, AcroSeal™

Catalog No. p-2799200
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 16853-85-3 | AlH4Li | 37.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Aluminum hydride
Color Colorless
Name Note 4.0M Solution in Diethyl Ether
CAS 16853-85-3, 60-29-7
Molecular Formula AlH4Li
Linear Formula LiAIH4
MDL Number MFCD00011075
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Density 0.7100g/mL
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
Solubility Information Solubility in water: vigorous reaction.
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES [Li+].[AlH4-]
IUPAC Name lithium(1+) alumanuide
Molecular Weight (g/mol) 37.95
PubChem CID 21226445
Formula Weight 37.95
Percent Purity 18.0 to 22.0% (as LiAlH4)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.71
Quantity 100 mL
Flash Point −017°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.