Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminum hydride, 4% in THF, Thermo Scientific™

Catalog No. AA45501AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45501AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45501AP Supplier Thermo Scientific Chemicals Supplier No. 45501AP

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-85-3 , 109-99-9
Molecular Formula AlH4Li
Molecular Weight (g/mol) 37.95
MDL Number MFCD00011075
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445
IUPAC Name lithium(1+) alumanuide
SMILES [Li+].[AlH4-]

Specifications

Color Gray
Physical Form Liquid
Quantity 500 mL
Solubility Information It reacts with water.
Formula Weight 37.95
Concentration or Composition (by Analyte or Components) Aluminum lithium hydride: 4%; Tetrahydrofuran: 96%
Ignition Point 321°C (610°F)
Sensitivity Air and moisture sensitive
Vapor Pressure 150mm Hg at 20°C
Density 0.9
Chemical Name or Material Lithium aluminum hydride
Show More Show Less
Hazard Category H225-H260-H302-H315-H318-H335-H336-H351
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334
DOTInformation Hazard Class: 4.3; Packaging Group: I
EINECSNumber 240-877-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.