Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminum hydride, 95% min

Catalog No. AA1344222
Click to view available options
Quantity:
25 g
100 g
500 g
5 x 25 g
5 x 100 g

Reducing Agent For Pharmaceuticals, Perfumes, and Fine organic Chemicals; Source of Hydrogen | CAS: 16853-85-3 | AlH4Li | 37.95 g/mol

Reducing agent for pharmaceuticals, perfumes and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizationsLithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It is also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-85-3
Molecular Formula AlH4Li
Molecular Weight (g/mol) 37.95
MDL Number MFCD00011075
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445
IUPAC Name lithium(1+) alumanuide
SMILES [Li+].[AlH4-]

Specifications

Melting Point >125°C (decomposition)
Color White to Gray
Physical Form Powder
Quantity 100 g
UN Number UN1410
Merck Index 14,349
Solubility Information Decomposes in water; Soluble in ether,diethyl ether and terahydrofuran.
Formula Weight 37.95
Percent Purity ≥95%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 0.917
Chemical Name or Material Lithium aluminum hydride
Show More Show Less
Hazard Category H260-H314-H500
Hazard Statement GHS H Statement
H260-H301-H314-H318;
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM ALUMINUM HYDRIDE
EINECSNumber 240-877-9
RTECSNumber BD0100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.