Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Lithium Aluminum Hydride Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
800 mL
4 x 100 mL
Description
Reactant or reagent for: The preparation of thermoplastic polyester polyamides from oleic acidLithium-polymer batteriesHydrodefluorination of gem-difluoromethylene derivativesAsymmetric aldol reactionsSynthesis of Li-Al-N-H composites with hydrogen absorption / desorption propertiesLAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcoholsSure/Seal is a trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 16853-85-3 |
| Density | 0.905 g/mL at 25°C |
| Molecular Formula | H4AlLi |
| Linear Formula | LiAlH4 |
| MDL Number | MFCD00011075 |
| Quantity | 25 mL |
| Synonym | LAH; Lithium alanate; Lithium tetrahydroaluminate |
| Molecular Weight (g/mol) | 37.95 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction