Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Bis(trifluoromethanesulfonimide), 99%

Catalog No. p-3212792
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 90076-65-6 | C2F6LiNO4S2 | 287.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90076-65-6
Molecular Formula C2F6LiNO4S2
Molecular Weight (g/mol) 287.09
MDL Number MFCD00210017
InChI Key QSZMZKBZAYQGRS-UHFFFAOYSA-N
Synonym lithium bis trifluoromethanesulfonyl imide, lithium bis trifluoromethyl sulfonyl amide, lithium bis trifluoromethanesulphonyl imide, litfsi, lithium bis trifluoromethylsulfonyl azanide, lithium bis trifluoromethylsulfonyl imide, bis trifluoromethane sulfonimide lithium salt, bistrifluoromethanesulfonimide lithium salt, lithium bis trifluoromethanesulfonyl amide, lithium triflimide
PubChem CID 3816071
IUPAC Name lithium;bis(trifluoromethylsulfonyl)azanide
SMILES [Li+].C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 234°C to 238°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: 10mg/mL (20°C)
Formula Weight 287.09
Percent Purity 99%
Physical Form Hygroscopic Powder
Chemical Name or Material Lithium bis(trifluoromethanesulfonimide)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.<
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.