Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™

Catalog No. p-7077198
Click to view available options
Quantity:
100 mL
800 mL
This item is not returnable. View return policy

Catalog No. AC209551000

Supplier: Thermo Scientific Chemicals 209551000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 4039-32-1 | C6H18LiNSi2 | 167.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium bis(trimethylsilyl)amide
CAS 4039-32-1, 109-99-9
Density 0.9000g/mL
Boiling Point 65.0°C
Flash Point −21°C
Molecular Formula C6H18LiNSi2
Linear Formula ((CH3)3Si)2NLi
Quantity 100 mL
Fieser 04,296; 05,393; 07,197; 12,280; 13,165; 14,194
Specific Gravity 0.9
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
Solubility Information Solubility in water: reacts.
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C
IUPAC Name lithium;bis(trimethylsilyl)azanide
Molecular Weight (g/mol) 167.33
PubChem CID 2733832
Formula Weight 167.33
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.