Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle

Catalog No. p-7046565
Click to view available options
Quantity:
100 mL
800 mL

CAS: 4039-32-1 | C6H18LiNSi2 | 167.327 g/mol

Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4039-32-1
Molecular Formula C6H18LiNSi2
Molecular Weight (g/mol) 167.327
MDL Number MFCD00008261
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
PubChem CID 2733832
IUPAC Name lithium;bis(trimethylsilyl)azanide
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C

Specifications

Quantity 100 mL
Linear Formula [(CH3)3Si]2NLi
UN Number UN2924
Beilstein 8136861
Flash Point −17°C (1°F)
Solubility Information Reacts with water.
Formula Weight 167.33
Concentration or Composition (by Analyte or Components) 20% (≈1.06M) soln. in THF/ethylbenzene
Sensitivity Air and moisture sensitive
Density 0.891
Chemical Name or Material Lithium bis(trimethylsilyl)amide, packaged in resealable septum cap bottle
Show More Show Less
Hazard Category H225-H302-H304-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 223-725-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.