Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Lithium Bis(Trimethylsilyl)Amide Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
800 mL
Description
Lithium bis(trimethylsilyl)amide solution (LiHMDS) is generally used in organic synthesis as a non-nucleophilic strong Bronsted base.Lithium bis(trimethylsilyl)amide is generally used in organic synthesis as a non-nucleophilic strong Br++nsted base. It can be used for salt metathesis reaction for the synthesis of cesium bis(trimethylsilyl)amide (CsHMDS) and lithium fluoride by reacting with cesium fluoride.
Specifications
Specifications
| CAS | 4039-32-1 |
| Density | 0.860 g/mL at 25°C |
| Molecular Formula | C6H18LiNSi2 |
| Linear Formula | [(CH3)3Si]2NLi |
| MDL Number | MFCD00008261 |
| Quantity | 100 mL |
| Synonym | LiHMDS; Hexamethyldisilazane lithium salt |
| Molecular Weight (g/mol) | 167.33 g/mol |
| Biological Activity | Central Nervous System |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction